ContactUsLinkComponent
RGB Color Analyzer
RGB Color Analyzer
Catalog # 470315-056
Supplier:  Avantor
undefined
RGB Color Analyzer
Catalog # 470315-056
Supplier:  Avantor
Specifications
  • Description:
    Color Analyzer
  • Cat. No.:
    470315-056
Specifications
About this item
The model H500 is an RGB color analyzer that can measure the amount of red, blue and green as well as the hue, saturation and luminance of items.
  • Measure both RGB and HSL levels
  • Relative (REL) feature allows the comparison of two-color samples
  • Easy to use
  • Two-year warranty