Adhesive Vinyl Labels 'Hazardous Waste Pickup and Accumulation', HCL Labels
Catalog # 103727-796
Supplier: HCL Label
Specifications
- Description:Preprinted adhesive labels
- Dimensions:6×11"
- Material:Vinyl, adhesive
- Cat. No.:103727-796
- Legend:Hazardous Waste Pickup and Accumulation
Specifications
About this item
HCL's multiple pickup hazardous waste label is great for documenting multiple pick-up dates without needing to apply a new label.
- Made of adhesive vinyl
This label combines the California and Federal Hazardous Waste label with the accumulation pick up record label. It's a great label to keep continuous records of multiple waste pick ups.
Certifications: California and Federal Hazardous Waste Compliant. Updated to meet the new 49 CFR 172.301 marking requirements.
Packaging: Supplied in packs of 25.