Adhesive Vinyl Labels 'No Pacemakers Allowed', HCL Labels
Catalog # 103727-924
Supplier: HCL Label
Specifications
- Description:Preprinted adhesive labels
- Dimensions:2×2"
- Material:Vinyl, adhesive
- Cat. No.:103727-924
- Legend:No Pacemakers Allowed
Specifications
About this item
No pacemakers graphic label is durable and easy to spot.
- Made of adhesive vinyl
- Bright red and black coloring for easy visibility.
Use this label in areas, or on equipment that should not be used by personnel with pacemakers.