Adhesive Vinyl Labels 'Caution This Tool', HCL Labels
Catalog # 103727-890
Supplier: HCL Label
Specifications
- Description:Preprinted adhesive labels
- Dimensions:8.5×11"
- Material:Vinyl, adhesive
- Cat. No.:103727-890
- Legend:Caution This Tool …
Specifications
About this item
Caution this tool label is made of adhesive vinyl and comes cleaned, double-bagged and ready for cleanroom usage.
- Cleaned, Double-bagged and ready for cleanroom usage
Use this label in cleanrooms to identify tools that are ready for decon, harvest, re-use or other specifications. This is a write-in label.
Packaging: Supplied in packs of 100.