Adhesive Vinyl Labels 'Caution: X-Ray Producing Machine -Write In', HCL Labels
Catalog # 103458-962
Supplier: HCL Label
Specifications
- Description:Preprinted adhesive labels
- Dimensions:3×5"
- Material:Vinyl, adhesive
- Cat. No.:103458-962
- Legend:Caution: X-Ray Producing Machine -Write In
Specifications
About this item
HCL's caution X-ray producing machine is made using a durable adhesive vinyl and matte lamination for extended durability.
Use this caution label to mark X-ray producing machines. Labels are laminated with a matte lamination in order to allow for easy write-ability. Label includes space for the building, room, manufacturer, model and serial number.
Packaging: Available in packs of 25.