Adhesive Vinyl Labels 'Danger - Hot Surface', HCL Labels
103727-860
- Description:Preprinted adhesive labels
- Dimensions:3×5"
- Material:Vinyl, adhesive
- Cat. No.:103727-860
- Legend:Danger - Hot Surface
Danger hot surface labels is printed on a durable adhesive vinyl and laminated for extended durability.
Use this danger label to identify hot surfaces that should not be touched. Surface needs to cool before performing any maintenance.
: ANSI Compliant graphics and header.