ContactUsLinkComponent
Professional Brawny Industrial® Medium Duty Premium DRC Wipers, Georgia Pacific®
Professional Brawny Industrial® Medium Duty Premium DRC Wipers, Georgia Pacific®
Catalog # 10714-054
undefined
Professional Brawny Industrial® Medium Duty Premium DRC Wipers, Georgia Pacific®
Catalog # 10714-054
Supplier Number:  GPC2007003CT
Specifications
  • Color:
    White
  • Length:
    16.3"
  • Material:
    Double recrepe (DRC)
  • Environmentally Preferable:
  • Cat. No.:
    10714-054
  • Packaging:
    Box
  • Supplier no.:
    GPC200-70/03
  • No. of towels/wipes per container:
    90
  • Ply:
    1
Specifications
About this item
Medium duty premium DRC wipers are strong, versatile wipers with excellent bulk, softness and absorption.
  • Cleans both water- and oil-based spills
  • Both general purpose and lab use
  • Performs like a rag without the expense
  • Tall box for easy dispensing
  • Works on dust, lint and liquid
  • Unscented
Dispenser box provides easy, one-handed, one-at-a-time dispensing.