Order Entry
United States
ContactUsLinkComponent
GOJO® TFX™ Green Certified™ Foam Soap Refill
GOJO® TFX™ Green Certified™ Foam Soap Refill
Catalog # 10713-970
undefined
GOJO® TFX™ Green Certified™ Foam Soap Refill
Catalog # 10713-970
Supplier Number:  GOJ566502CT

Specifications

  • Capacity:
    1200 ml (40.6 oz.)
  • Color:
    Yellow
  • Description:
    foam hand cleaner refill, unscented
  • Cat. No.:
    10713-970
  • For use with:
    Refill for TFX® Touch-Free Automatic Foam Soap Dispenser
  • Supplier no.:
    GOJ5665-02

Specifications

About this item

Refill for TFX® Touch-Free Automatic Foam Soap Dispenser (sold separately).

  • Mild foaming soap formula certified by Green Seal and EcoLogo
  • For use on hands
Certifications: EcoLogo Certified, Green Seal Certified.