ContactUsLinkComponent
Rubbermaid® Commercial Stacking Recycle Bin
  10158-706
undefined
Rubbermaid® Commercial Stacking Recycle Bin
  10158-706
 :  RCP571473BE
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Depth:
    21"
  • Color:
    Blue
  • Height:
    14¾"
  • Cat. No.:
    10158-706
  • Shape:
    Rectangular Prism
  • Supplier no.:
    5714-73BLU
The Rubbermaid Recycling Bin is made from linear low density polyethylene to withstand harsh weather conditions and prevent cracking, denting, or bowing.
  • For indoor recycling
  • Stackable for convenient use of multiple containers at one time
  • Nestable for easy storage and shipping
  • Texture and design prevent liquid and debris build up for easy cleaning
  • Rigid plastic liner material
  • Open top
Inboard handles for easy lifting and carrying, meeting municipality container requirements.