ContactUsLinkComponent
Bone Clones® Fossil Hominid Tools set of 6
Bone Clones® Fossil Hominid Tools set of 6
Catalog # 470317-812
undefined
Bone Clones® Fossil Hominid Tools set of 6
Catalog # 470317-812
Supplier Number:  MS-99-SET
Specifications
  • Description:
    Fossil Hominid Tools set of 6
  • Cat. No.:
    470317-812
Specifications
About this item
Set of 6 fossil hominid tools.
  • Casting from originals that present diverse osteological characteristics, each Bone Clones® cast is hand-made from start to finish
  • Rigorous standards are applied when selecting original specimens to ensure the best representation
  • Proprietary, custom blend of polyurethane developed to capture and replicate minute detail
ncludes an Acheulian Banded Flint Hand-Axe from southwest Morocco; a Mousterian Neanderthal Triangular Bi-facial Hand-Axe; a Mousterian Bi-facial ovate point on a Levallois Blade; a Mid-Acheulian Cordiform Bi-facial Hand-Axe; a Mousterian 40,000 year old scraper; and an Acheulian tool. Acheulean tools (approximately 2.0 MYA-200,000 YA) are associated with Homo erectus. Mousterian tools are dated at (100,000-40,000 YA).
Weight: 2 lbs.
Accessories information: Accessories not included.