Specifications
- Description:Sahelanthropus tchadensis Cranium
- Size:7D×4W×4"H
- Cat. No.:470317-800
- Weight:1 lb.
Specifications
About this item
6-7 MYA. The Sahelanthropus tchadensis skull was discovered by Michael Brunet's team in Chad in 2001 and described in Nature in 2002.
- Each Bone Clones® cast is hand-made from start to finish
- Rigorous standards are applied when selecting original specimens to ensure the best representation
- Proprietary, custom blend of polyurethane developed to capture and replicate minute detail
Some suggest that S. tchadensis existed near the time that hominids and apes separated on their evolutionary paths. It could be that this specimen is a representative of an early hominid, predating A. afarensis by 3 to 4 million years; on the other hand, it might be an ancestor of the gorilla. The characteristics of the cranium are a mosaic of hominid-like (short face, the size and shape of the canines), and ape-like (very large browridges and small brain case) features.
The Bone Clones® Hominid line is composed of discoveries from anatomically modern humans, archaic humans, early Homo, early hominins, and other hominids. The majority of the casts in this line have been recreated by a team of anatomical sculptors. Some are reconstructions made by anthropology professionals using fragmentary elements from original discoveries and extrapolating the missing parts from those.