ContactUsLinkComponent
Corepoint Scientific™ Hazardous Location Refrigerators and Freezers, Horizon Scientific
undefined
Corepoint Scientific™ Hazardous Location Refrigerators and Freezers, Horizon Scientific
  • Catalog No:
  • 76335-206
  • 76335-204
  • 76335-196
  • Environmentally Preferable:
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Cat. No.:
  • 76335-206
  • 76335-204
  • 76335-196
  • Defrost:
  • Manual
  • Cycle
  • Manual
  • Shelves/drawers:
  • 3 adjustable shelves (refrigerator)
  • 5 adjustable shelves
  • 4 fixed shelves
  • Door Type:
  • 1 solid swing door, hinge right, 1 interior door (freezer)
  • 1 solid swing door, hinge right
  • 1 solid swing door, hinge right
  • Material:
  • Steel
  • Steel
  • Steel
  • Electrical:
  • 115 V, 6 A
  • 115 V, 6 A
  • 115 V, 6 A
  • Weight:
  • 185.1 kg (408 lbs.)
  • 178.3 kg (393 lbs.)
  • 176.9 kg (390 lbs.)
  • Environmental Certification:
  • HFC Refrigerant
  • HFC Refrigerant
  • HFC Refrigerant
Frequently Bought Together