Order Entry
United States
ContactUsLinkComponent
 
Rubbermaid® Commercial Slim Jim® Bottle and Can Recycling Top
  10158-634
undefined
Rubbermaid® Commercial Slim Jim® Bottle and Can Recycling Top
  10158-634
 :  RCP269288GN

 

  • Depth:
    11⅜"
  • Color:
    Green
  • Height:
    2¾"
  • Material:
    Plastic
  • Cat. No.:
    10158-634
  • Shape:
    Rectangular
  • For use with:
    Rubbermaid® Commercial Slim Jim® Containers
  • Supplier no.:
    2692-88GRE
  • Lid type:
    Bottle and can recycling top

 

 

The Rubbermaid Commercial Untouchable Bottle and Can Recycle Bin Top is made from high density polyethylene, which resists moisture and chemicals.

  • Fits your Rubbermaid Commercial Untouchable square recycling bins
  • Ultraviolet stabilizer minimizes discoloring and fading
  • High-density polyethylene is strong and resists moisture
  • Color-coded tops encourage separation of recyclables.

The rectangular top has two round openings for easily depositing recyclable bottles and cans.