To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Depth:19½"
- Color:Blue
- Height:27⅝"
- Material:Plastic
- Cat. No.:10158-588
- Shape:Rectangular Prism
- Supplier no.:3958-73BLU
Rubbermaid commercial square recycle bins are perfect for use in areas of high paper generation, such as near copiers, printers, and in mailrooms.
- Perfect for high volume paper generation
- Contains post-consumer recycled resin
- Exceeds EPA guidelines
- Designed to withstand extreme weather
- open top
This recycle bin contains post-consumer recycled resin (PCR), exceeding EPA guidelines. The recycling bin station containers serve as a convenient central collection site for multiple workstations. Recommended for indoor and outdoor use. The large capacity is ideal for heavy traffic areas. These recycle bins are designed to withstand extreme weather and handling.