Mercury Floor Machines, PRO-175, Essendant LLC FKA Lagasse
Supplier: Janitorial Supplies
Certificates
About this item
Precision-balanced, heavy-duty 66-frame motor ensures reliable, professional-quality performance.
- Triple-planetary, high-torque transmission
- Safety-enhanced interlock switch on the handle leaves both hands conveniently in control
- Handle is fully adjustable and is comfortable for all operators
- 50' cord length
- 175 rpm (top speed)
- 1.5 horsepower
- 16" brush diameter
Triple-planetary, high-torque transmission stands up to tough tasks, while minimizing gear wear. Safety-enhanced, interlocking auto-cutoff switch enables both left- and right-hand control. Nonmarring, full-wrap bumpers protect walls and baseboards from scuffs and scrapes. Fully adjustable handle accommodates nearly any user. Smooth-rolling 6" wheels simplify transport.
Certifications: UL listed.
Ordering information: Pads and pad drivers (Sold Separately).
Frequently Bought Together