Water with 0,1% (v/v) acetic acid, LiChrosolv®, hypergrade for LC-MS, Supelco®
Catalog # EM1.59007.2500
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:2.5 L
- Cat. No.:EM1.59007.2500
Specifications
About this item
:
Boiling Pt:100 °C (1013 hPa)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
:7732-18-5