Impeller Set (Shaft and Blade Kit) for Model BDC250, Caframo
Catalog # 76019-362
Supplier: Caframo
Specifications
- Description:Overhead Stirrer Accessory
- Dimensions:Length: 73/8", Ø 0.24"
- Cat. No.:76019-362
- Blade size:0.4×1.1"
Specifications
About this item
Impeller set contains two blades and shaft. Includes axial and radial 0.4×1.1" blades 73/8×0.24" shaft and hardware 304 stainless steel.
- Electropolished 316L stainless steel – Chemical and corrosion resistant
- Axial and radial blades included – Choice of two types of flow
- Set screw compatible with standard tools – Blades easily interchangeable
Shaft and impeller blades to fit collet of BDC250 (97001-138) stirrer, all electropolished 316L stainless steel. Includes one radial blade and one axial blade and interchangeable set screw.
Ordering information: This kit is included when a BDC250 is purchased.