Order Entry
United States
Orders LinkContactUsLinkComponent
Hematoxylin solution (Harris) mercury-free

Hematoxylin solution (Harris) mercury-free

500 ml
Catalog # 101410-908
Health
Supplier #:  26041-20
Hematoxylin solution (Harris) mercury-free
Catalog # 101410-908
CAS Number:  517-28-2
Hematoxylin solution (Harris) mercury-free
Hematoxylin solution (Harris) mercury-free

500 ml

CAS Number:  517-28-2
Price
$75.44
Log in for availability
Each (500ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Stain selectivity for nuclei, fat and lipids. Ready-to-use.

Synonyms:  DcH-48, Natural Black I, Hydroxybrasilin
  • CAS Number:517-28-2
  • Formula:C₁₆H₁₄O₆
  • MDL Number:MFCD00078111
  • Molecular Weight:302.28 g/mol
Specifications

Specifications

  • Size:
    500 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.