Orders LinkContactUsLinkComponent
Digital Multimeter E
Digital Multimeter E
Multimeter E
Catalog # 470316-734
Supplier #:  1018832
Digital Multimeter E
Catalog # 470316-734
Digital Multimeter E
Digital Multimeter E
Multimeter E
Price
$61.39
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Compact 31/2digit multimeter for measuring voltage, current and resistance as well as for diode and hFE gain testing. All measurement ranges are selected by means of a rotary dial. All measurement ranges are protected against overload.
Ordering information: Includes measuring leads and battery.
Specifications
  • Description:
    Multimeter E