Phenol Red Dextrose Broth
Supplier: Ward's Science
Jump to:
About this item
To prepare media, add 20 g of phenol red dextrose broth to a liter of distilled or de-ionized water. The media will not be sterile until autoclaved prior to use.
2 Options Available Below
Product Details & Documents
To prepare media, add 20 g of phenol red dextrose broth to a liter of distilled or de-ionized water. The media will not be sterile until autoclaved prior to use.
Product Family Options
Product Information
- SizeAvailabilityPrice
- Supplier Number: AD-MM1038-100G
- Supplier Number: AD-MM1038-500G