About this item
Ward's® pure preserved specimens-the best dissection experience from start to finish.
- Safest specimens
- Low formalin odor
- Best classroom experience
Dissection specimens that encourages discovery and real life scenarios. Learn areas of the brain, discuss ramifications of the tumor or stroke in your patient, and find simulated tumors and/or stroke in this all new interactive product.
Sheep brains contain simulated tumors in three different areas of the brain: the hypothalamus, motor cortex, and visual cortex. Sheep brains contain simulated strokes in either the motor cortex or the visual cortex. Each set comes with either one, two, five or ten of each example.
Hypothalamus: The hypothalamus main role is to keep the body in homeostasis as much as possible. Examples include regulating body temperature, emotions, blood pressure and heart rate, appetite.
Occipital lobe: Controls visual functions.
Motor cortex: Involved in planning, control, and execution of voluntary movements.