Orders LinkContactUsLinkComponent
Ward's® Essentials Research Binocular Microscopes
 :   VWR
Ward's® Essentials Research Binocular Microscopes
 :  VWR
Ward's® Essentials Research Binocular Microscopes
Ward's® Essentials Research Binocular Microscopes
Packed with standard research features, these microscopes are ideal for educational and professional applications.
  • Professional Siedentoph binocular head
  • Six magnification levels: 40×, 100×, 250×, 400×, 1000× and 2500×
  • 10× and 25× interchangeable wide-field eyepieces
  • 1 year limited warranty
Combining an ergonomic design and advanced features this microscope fits the needs of student and clinical environments. Ultra-precise focusing and built-in mechanical stage contribute to the ease of use and the glass optics and cool adjustable LED illumination provide clear, crisp images.
The 1.3MP USB camera includes user-friendly editing and processing software compatible with Windows XP, Vista, 7, 8 and 10.
Product Family Options
  • Description
    Illumination
  • Compound Binocular Microscope
    LED
    Each
    Each - $354.15
    $354.15
    Each
       $354.15
     : SW380B
  • Compound Binocular Microscope with USB Camera
    LED
    Each
    Each - $424.21
    $424.21
    Each
       $424.21
     : SW380B+EP1