MicroMax® NS Disposable Shoe Covers with Vinyl Sole, Lakeland Industries
Supplier: Lakeland
Certificates
About this item
Disposable protective shoe covers with non-skid vinyl sole are made from lightweight microporous fabric for protection from bloodborne pathogens.
- Microporous film laminated to polypropylene substrate
- Serged seams
- Blood-borne pathogen/viral barrier tested
- Elastic top
These covers provide the protection needed from dirt, dust, grease, grime, and light chemical splash, as well as blood/viral protection. Covers feature vinyl sole for use on wet surfaces.
Frequently Bought Together