TRITON™ X-114 (Polyethylene glycol tert-octylphenyl ether) solution 10%
Supplier: Avantor
Certificates
About this item
CAS Number:9036-19-5
MDL Number:MFCD00128254
Storage Temperature:Refrigerator
UN:3082
ADR:9,III
:GHS05
:GHS07
:GHS09
Frequently Bought Together