Test Tube ATC pH Pen, Sper Scientific
Catalog # 10852-662
Supplier: Sper Scientific
Specifications
- Description:ATC pH Pen for Test Tubes
- Compliance:IP65
- Dimensions:225×35×20 mm (9×13/8×1")
- Sensor type:pH probe (5⅛" × ½" (130 × 12 mm))
- Temperature Compensation:Automatic
- Weight:66 g (2.3 oz.)
- pH Accuracy:±0.1
- pH Calibration:3 points
- pH Range:0 to 14
- pH Resolution:0.01
- Cat. No.:10852-662
- Supplier no.:850063
Specifications
About this item
Fits standard test tubes 16x50 mm or larger.
- Features hold function
- Auto power off with low battery indicator
- Highly stable and accurate readings with a large LCD display
- Designed for simple one-hand operation
- Automatic temperature compensation
Automatically calibrates to pH 4, 7 and 10 buffers and can be adjusted to recognize some other non-standard buffers as well.
Certifications: The case is IP65 waterproof and will float if accidentally dropped into the water.
Delivery: Comes with two button-cell batteries and a built-in electrode kept moist with a protective cap and included solution.