Test Tube pH Pen, Sper Scientific
10852-660
- Description:Test Tube pH Pen
- Compliance:IP65
- Dimensions:225×35×20 mm (9×13/8×1")
- Sensor type:pH probe (5⅛" × ½" (130 × 12 mm))
- Weight:66 g (2.3 oz.)
- pH Accuracy:±0.2
- pH Calibration:3 points
- pH Range:0 to 14
- pH Resolution:0.01
- Cat. No.:10852-660
- Supplier no.:850062
Fits standard test tubes 16×50 mm or larger.
- Designed for simple one-hand operation
- Highly stable and accurate readings with a large LCD display
- Hold function, and auto power off with low battery indicator
Automatically calibrates to pH 4, 7 and 10 buffers and can be adjusted to recognize some other non-standard buffers as well.
: The case is IP 65 waterproof and will float if accidentally dropped into the water.
: Comes with two button-cell batteries and a built-in electrode kept moist with a protective cap and included solution.