ContactUsLinkComponent
Tech-Med® Wartenberg Pinwheels, Dukal
Tech-Med® Wartenberg Pinwheels, Dukal
Catalog # 76236-498
Supplier:  Dukal
undefined
Tech-Med® Wartenberg Pinwheels, Dukal
Catalog # 76236-498
Supplier:  Dukal
Supplier Number:  7020
Specifications
  • Description:
    Tech-Med® Wartenberg Pinwheel
  • Cat. No.:
    76236-498
Specifications
About this item
Also known as a Wartenberg Neurowheel, the instrument is designed to test nerve reactions (sensitivity) as it rolled systematically across the skin.
  • Chrome finish
  • 7.5" overall height