ADC® E-Sphyg Digital Pocket Aneroid Sphygmomanometer
Catalog # 470315-696
Supplier: AMERICAN DIAGNOSTIC CORP CP
Specifications
- Color:Black
- Description:E-Sphyg
- Size:Adult (23 - 40 cm)
- Cat. No.:470315-696
Specifications
About this item
Definitely not a electronic home monitor.
- Extra large LCD screen
- Backlit display
- Auto off function
- Works with most cuffs
- Not made with natural rubber latex
ADC®, a world leader in diagnostic products, has completely reinvented the aneroid to produce an instrument that combines precision crafted performance, rugged durability, and incredible ergonomics. It has replaced the diaphragm, gears, springs, and linkages - the very components that make an aneroid vulnerable to shock, vibration, over-inflation and wear - and replaced them with electronic pressure sensors, IC chips and a backlit extra large, easy to read, liquid crystal digital display.