ADC® Adscope® 658 Electronic Stethoscopes
Supplier: Avantor
Certificates
About this item
Hear sounds you may have missed with an amplified stethoscope.
- Amplification of up to 24 db
- Eight step volume control
- Noise filtering contact microphone
- High impact housing
- Not made with natural rubber latex
With the ADC® adscope 658 electronic stethoscope, helpful when auscultating faint heart and lung sounds, when the patient is obese, or when listening through clothing. The bi-lumen (dual channel) cariology headset optimizes the acoustic pathway, with heavy walled tubing, large-diameter aural tubes, and a double-leaf tensioning spring that with stands thousands of cycles.