ContactUsLinkComponent
Low-Profile Common Ground Cord, SCS, Desco Industries
Low-Profile Common Ground Cord, SCS, Desco Industries
Catalog # 89024-542
Supplier:  Desco
undefined
Low-Profile Common Ground Cord, SCS, Desco Industries
Catalog # 89024-542
Supplier:  Desco
Supplier Number:  LPCGC151M
Specifications
  • Description:
    Low-Profile Common Ground Cord
  • Length:
    15'
  • Cat. No.:
    89024-542
  • Supplier no.:
    LPCGC151M
Specifications
About this item
With a low-profile top design it reduces snagging of clothes and work instruments, this common point ground cord connects two wrist straps and a work surface to ground, helping ensure that all parts are at the same ground potential.
  • 15 foot ground cord with 10 mm stud snap
  • Two banana jacks provide two grounding points for wrist straps or carts
  • 10 ring terminal at end of ground cord allows easy attachment to electrical ground
  • Includes 1 megohm resistor
  • Made in the United States of America
Easy installation requires simply snapping cord to a mat or work surface. Plug in the wrist strap coil cord. Equipped with holes for permanent mounting to a bench.
Certifications: Meets ANSI/ESD S20.20 and Grounding standard ANSI/ESD S6.1.