Standard Red Elastic Heel Grounder, SCS, Desco Industries
Catalog # 89024-526
Supplier: Desco
Specifications
- Description:Standard Red Elastic Heel Grounder
- Cat. No.:89024-526
- Supplier no.:HGS1MD-RED
Specifications
About this item
Standard heel grounder features a D-ring strap and elastic hook and loop closure for adjustability.
- Compliance verification is simplified by the bright red elastic strap
Heel Grounders are designed and manufactured for comfort, durability and cost effectiveness.
Certifications: ANSI/ESD S20.20, EIA 625, ESD-SP9.2.