Order Entry
United States
ContactUsLinkComponent
 
All-Purpose Spill Clean Up Shaker, Spill Magic, Acme United
Catalog # 76327-126
Supplier:  First Aid Only
undefined
All-Purpose Spill Clean Up Shaker, Spill Magic, Acme United
Catalog # 76327-126
Supplier:  First Aid Only
Supplier Number:  97504

Specifications

  • Description:
    Absorbent Powder
  • Height:
    8.5"
  • Length:
    3.25"
  • Size:
    6/pack
  • Cat. No.:
    76327-126
  • Packaging:
    Shaker bottle
  • Form:
    Powder
  • Weight:
    145 g
  • Width:
    3.23"

Specifications

About this item

Spill magic lightweight, high performance, all-purpose absorbent powder removes a liquid or semi-liquid spill of any source or viscosity almost instantly.

  • 25 oz. shaker of lemon scented absorbent‚powder
  • Shaker for easy dispersal of powder on spill
  • Quick and easy to use
  • Non-toxic
  • Absorbs spill instantly leaving the surface clean & dry
  • For any spill (except Hydrofluoric Acid)
  • For use on asphalt, tile or concrete surfaces

Spill magic is a quick clean up solution that leaves the surface dry and residue free. Save time, money and reduce slip and fall instances.

The powder is ideal for cleaning industrial, food or biohazard spills. The portable shaker bottle allows for easy dispersal of the powder around the spill.

Accessories information: Accessories not included.
Caution: Used Spill Magic must be disposed of in accordance with any federal, state or local laws and regulations governing the absorbed liquid or material. Do not use to absorb hydrofluoric acid. Do not pour down drain.