Organic Chemistry Set
Catalog # 470313-394
Supplier: GSC International
Specifications
- Description:Organic Chemistry Set
- Cat. No.:470313-394
Specifications
About this item
This set is made from high-quality borosilicate glass and all joints are ground 19/22.
Set Includes: (1) West condenser, (1) Distilling Column, (5) Round-bottom flask (500mm, 250mm, 100mm, 50mm, 25mm), (1) Separatory funnel, 100ml, (1) Bleed tube, (1) Thermometer holder, (1) Thermometer adapter, (1) Three-way adapter, (1) Vacuum adapter, (1) Claisen adapter, (1) Stopper.