Order Entry
United States
ContactUsLinkComponent
SterEM™ Tryptic Soy Agar (TSA), USP , Irradiated, Triple Bagged, Red Tinted
SterEM™ Tryptic Soy Agar (TSA), USP , Irradiated, Triple Bagged, Red Tinted
  76200-296
 :  
undefined
SterEM™ Tryptic Soy Agar (TSA), USP , Irradiated, Triple Bagged, Red Tinted
  76200-296
 :  W540R
 :  

 

  • Description:
    Tryptic Soy Agar (TSA), SterEM™, irradiated, USP, triple bagged, red tinted
  • Dimensions:
    15×100 mm (Plate)
  • Cat. No.:
    76200-296
  • pH:
    7.3 ±0.2 at 25 °C
  • Fill volume:
    34 ml
  • Storage temperature:
    15...30 °C
  • Application:
    Cosmetics, compounding pharmacy, pharmaceutical manufacturing

 

 

Sterile plates for environmental monitoring in a clean room or clean bench setting.

  • Room temperature storage
  • Improved moisture control packaging format
  • Optically clear polystyrene - For distortion free viewing
  • Heavy-Duty - Reduces breakage during shipping
  • Stacking rings - Polystyrene Petri plates feature raised rings for stacking ability and prevents sliding
  • Sterile by Irradiation - Validated for sterility assurance level (SAL) of 10⁻⁶
  • Triple Bagged - Two nylon inner bags. One outer Mylar(R) bag that is heat sealed
  • Outer bag is VHP impermeable
  • Red Tinted plate - Easy visual ID enables easy differentiation between TSA and Sabdex

Irradiated Tryptic Soy Agar (TSA), USP, is recommended for use as a general growth medium for the detection and enumeration of microorganisms. The plates are triple bagged and sterilized by irradiation to promote a higher sterility assurance level.

 : Manufactured in a licensed cGMP facility with a quality management system certified to ISO 13485.