NEW AGE® Vinyl Powder-Free Disposable Gloves, Tronex®
: Tronex
Tronex® vinyl powder-free gloves are food grade safe and offer excellent tactile sensitivity.
- Proprietary material formulation designed for outstanding performance integrity, elasticity, and comfort
- Not made with natural rubber latex
- Conforms to the hands, providing exceptional tactile sensitivity and softness for reduced hand fatigue
- Ambidextrous with beaded cuffs
- Food grade safe
- Powder-free, smooth finish
Proprietary polyvinyl chloride (PVC) material formulation is designed for outstanding performance integrity, elasticity, and comfort. An excellent alternative to latex, these gloves are ideal for various foodservice, catering, and light food processing applications. Available in natural or blue.
: Gloves are packaged 100 per box, 10 boxes per case.
- Catalog No:
- 76208-522
- 76208-524
- 76208-520
- 76208-526
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable/Reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-Free
- Latex-Free
- Latex-Free
- Latex-Free
- Powdered/Powder-free:
- Powder-Free
- Powder-Free
- Powder-Free
- Powder-Free
- Finish:
- Smooth
- Smooth
- Smooth
- Smooth
- Cuff Style:
- Beaded
- Beaded
- Beaded
- Beaded
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Food Contact Approved:
- +
- +
- +
- +
Frequently Bought Together