Order Entry
United States
ContactUsLinkComponent
Novagen® pET-28a(+) DNA, MilliporeSigma
Novagen® pET-28a(+) DNA, MilliporeSigma
Catalog # EM69864-3
Supplier:  MilliporeSigma
CAS Number:  
Novagen® pET-28a(+) DNA, MilliporeSigma
Catalog # EM69864-3
Supplier:  MilliporeSigma
Supplier Number:  69864-3
CAS Number:  

Specifications

  • Description:
    pET-28a(+) DNA
  • Size:
    10 μg
  • Storage and Handling:
    Standard Handling
  • Storage Temperature:
    ≤–70 °C
  • Cat. No.:
    EM69864-3
  • Supplier no.:
    69864-3

Specifications

About this item

The pET-28a-c(+) vectors carry an N-terminal His Tag/thrombin/T7 Tag configuration plus an optional C-terminal His Tag sequence.

  • Sequence is numbered by the pBR322 convention
  • T7 expression region is reversed on the vector map (TB074)

The pET Vectors are supplied as purified plasmid DNA (10 µg). Each order of pET DNA also includes an Induction Control strain (supplied as a glycerol stock).