Vinyl Powder-Free Disposable Gloves, Tronex®
Supplier: Tronex
Certificates
About this item
Tronex® vinyl powder-free gloves are food grade safe and offer excellent tactile sensitivity.
- Latest generation material formulation offering enhanced comfort and lightness of fit
- Not made with natural rubber latex
- Contoured fit with excellent tactile sensitivity for reduced hand fatigue
- Ambidextrous with beaded cuffs
- Ideal for frequent change environments
- Powder-free, smooth finish
Latest generation polyvinyl chloride (PVC) material formulation is an excellent alternative to latex. Ideal for use in frequent change environments, as well as food handling, prepartion, and serving.
Packaging: Gloves are packaged 150 per box, 10 boxes per case.
Specifications
- Catalog No:
- 76208-518
- 76208-514
- 76208-516
- 76208-012
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable/Reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-Free
- Latex-Free
- Latex-Free
- Latex-Free
- Powdered/Powder-free:
- Powder-Free
- Powder-Free
- Powder-Free
- Powder-Free
- Finish:
- Smooth
- Smooth
- Smooth
- Smooth
- Cuff Style:
- Beaded
- Beaded
- Beaded
- Beaded
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile