ContactUsLinkComponent
Ward's® Pure Preserved Pregnant Sheep Uterus
Ward's® Pure Preserved Pregnant Sheep Uterus
  470314-612
 :  Avantor
undefined
Ward's® Pure Preserved Pregnant Sheep Uterus
  470314-612
 :  Avantor
  • Description:
    Pure Sheep Uterus Pregnant
  • Injection type:
    Uninjected
  • Cat. No.:
    470314-612
  • Packaging:
    Pail of 1
Ward's® PURE Preserved Specimens - the best dissection experience from start to finish.
  • Safest specimens
  • Low formalin odor
  • Best classroom experience
Ward's® PURE preserved specimens offer the best classroom experience because PURE specimens are safe. Students stay focused on learning and not on smells. Experience quality with true-to-life preservation, combined with artfully illustrated dissection guides, free with every purchase, to maximize value, lesson time and student engagement.
We guarantee you’ve never experienced dissection like this before. Go Pure.
This specimen is an excellent supplement to our fetal lambs. Use both to allow students a view of the sheep’s initial developmental stages, as well as the female reproductive system. The size of fetus will vary depending on the gestation period provided.
 : Ward's® preserved sheep materials is a byproduct of the food industry. Sheep are not raised specifically for dissection. These animals are 'recycled' for educational use.