ZORBAX® RRHD - Small Molecule Columns, Agilent Technologies
About this item
ZORBAX® Rapid Resolution High Definition (RRHD) columns are an expansion of the ZORBAX 1.8 μm particle column line, available in 50, 100 and 150 mm lengths for fast or high resolution, truly high definition, separations of your most complex samples.
- High pressure (1200 bar) columns for optimum results with the 1290 Infinity LC or other UHPLC instruments
- 1.8 μm particles deliver maximum resolution for the most defined separations
- Achieve the same selectivity on 3.5 and 5 μm ZORBAX columns with the same bonded phase for compatibility with any LC
ZORBAX® RRHD columns are available in four wide pore 300 Å phases for bio separations, including ZORBAX RRHD diphenyl. Also available: ZORBAX® RRHD 300-HILIC columns for glycopeptide analysis, which enable to resolve peptide glycoforms, 12 ZORBAX® phases, including Eclipse Plus C18 for superior peak shape, ZORBAX® StableBond C18 for low pH stability, Bonus-RP, Eclipse PAH, Eclipse Plus Phenyl-Hexyl and Extend-C18, HILIC Plus.
0 Options Available Below
Product Details & Documents
ZORBAX® Rapid Resolution High Definition (RRHD) columns are an expansion of the ZORBAX 1.8 μm particle column line, available in 50, 100 and 150 mm lengths for fast or high resolution, truly high definition, separations of your most complex samples.
- High pressure (1200 bar) columns for optimum results with the 1290 Infinity LC or other UHPLC instruments
- 1.8 μm particles deliver maximum resolution for the most defined separations
- Achieve the same selectivity on 3.5 and 5 μm ZORBAX columns with the same bonded phase for compatibility with any LC
ZORBAX® RRHD columns are available in four wide pore 300 Å phases for bio separations, including ZORBAX RRHD diphenyl. Also available: ZORBAX® RRHD 300-HILIC columns for glycopeptide analysis, which enable to resolve peptide glycoforms, 12 ZORBAX® phases, including Eclipse Plus C18 for superior peak shape, ZORBAX® StableBond C18 for low pH stability, Bonus-RP, Eclipse PAH, Eclipse Plus Phenyl-Hexyl and Extend-C18, HILIC Plus.
Product Family Options