ContactUsLinkComponent
SmartPro UPS Battery Backup, Line-Interactive, Tripp Lite
SmartPro UPS Battery Backup, Line-Interactive, Tripp Lite
Catalog # 89079-136
Supplier:  Tripp Lite
SmartPro UPS Battery Backup, Line-Interactive, Tripp Lite
Catalog # 89079-136
Supplier:  Tripp Lite
Supplier Number:  SMART1000LCD
Specifications
  • Color:
    Black
  • Description:
    UPS Battery Backup
  • Cat. No.:
    89079-136
  • Supplier no.:
    SMART1000LCD
Specifications
About this item
The SmartPro LCD 120 V, 1000 VA/500 W line-interactive uninterruptible power supply provides battery backup and AC power protection against blackouts, brownouts, power surges and line noise that can damage electronics or destroy data.
  • Supports a basic desktop PC up to 60 min. during power outage
  • Features 8 protected outlets - 4 battery protected, 4 surge only
  • Offers automatic voltage regulation (AVR)
  • LCD screen reports real-time UPS and power status
  • Plugs into any NEMA 5-15R socket
  • Three years limited warranty
Ideal for backing up your desktop computer or audio/visual components, this line-interactive UPS switches to battery backup mode in milliseconds to keep your connected equipment running long enough to save files and shut down safely with no data loss.
Accessories information: HG outlet cover compatible with all NEMA 5-15R-HG and NEMA 5-15R outlets. Self locking feature allows for easy installation. Plugs are removable, reusable and designed for repeated use.