


Hemacolor® Rapid staining of blood smear solution I fixative reagent, Sigma-Aldrich®
Catalog # EM1.11955.2500
Supplier: MilliporeSigma
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Size:2.5 L (preparation capacity 1250 - 5000 slides)
- Cat. No.:EM1.11955.2500
- Supplier no.:1.11955.2500
Specifications
About this item
:
UN:1230
ADR:3 (6.1),
:GHS02
:GHS06
:GHS08
Storage Temperature:Ambient
:1