ContactUsLinkComponent
EZClick™ Palmitoylated Protein Assay Kit (FACS/Microscopy), Green Fluorescence, BioVision
EZClick™ Palmitoylated Protein Assay Kit (FACS/Microscopy), Green Fluorescence, BioVision
Catalog # 76326-214
Supplier:  Avantor
CAS Number:  
undefined
EZClick™ Palmitoylated Protein Assay Kit (FACS/Microscopy), Green Fluorescence, BioVision
Catalog # 76326-214
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Palmitoylated Protein Assay Kit (FACS/Microscopy), Green Fluorescence
  • Size:
    100 Assays
  • Environmentally Preferable:
  • Cat. No.:
    76326-214
Specifications
About this item
For labeling and detection of Palmitoylated proteins.
  • Highly specific
  • Robust method for labeling and detection of palmitoylated proteins
  • Simple procedure
Palmitoylation occurs when fatty acids, such as palmitic acid are covalently attached to the side chains of cysteine (and less frequently to serine, threonine side chains) in proteins. This type of post-translational modification greatly affects cellular trafficking, compartmentalization, and membrane tethering. Palmityltransferase (PAT), the enzyme responsible for this modification transfers a palmitate moiety from palmitoyl-CoA to the thiol group of cysteine in the target protein. Compared to myristoylation, palmitoylation is reversible and the reverse reaction is catalyzed by thioesterases. The Palmitoylation/Depalmitoylation cycle plays an important role when modified protein shuttles between cellular compartments. PAT mutations are associated with many neurological diseases and cancer progression. BioVision offers EZClick™ palmitoylated protein assay kit, a highly specific, simple and robust method for labeling and detection of palmitoylated proteins. The kit uses a modified palmitic acid that is fed directly into the cells and gets incorporated into proteins during or post translation. This post translational modification can be followed by click reaction with an azide-containing dye. The assay kit offers a powerful method for imaging localization, trafficking, and dynamics of palmitoylated proteins, or detection by FACS for quantitative studies. Labeled palmitoylated proteins can be directly detected in 1D or 2D gels using the appropriate excitation sources, or enriched by immunoprecipitation with biotin-azide or antibodies prior to proteomic analysis. We provide sufficient materials for 100 assays in a 96-well plate format.