5 L Process Filter Reactors, Cylindrical, Unjacketed, Electric, Chemglass
Catalog # 10170-694
Supplier: Chemglass
Small Business Enterprise
Specifications
- Description:5 l Process Filter Reactor, Unjacketed, Electric, Complete
- Cat. No.:10170-694
- Supplier no.:CG-1959-U5000
Specifications
About this item
Unjacketed filter reactors can be used for single or multi-step reactions and filtrations in the same vessel, and large scale peptide synthesis.
Filter base eliminates material transfers and significantly reducing losses. Complete reactor system on our ChemRxnHub stand. Supplied with the components listed in the link below. 150°C max. temp. limit for filter base.