Naphthol AS-E phosphate (KG-501) ≥98%
Supplier: Avantor
Certificates
About this item
Synonyms: 3-((4-Chlorophenyl)carbamoyl)naphthalen-2-yl dihydrogen phosphate, N-(4-Chlorophenyl)-3-(phosphonooxy)naphthalene-2-carboxamide
CAS Number:18228-17-6
:
Formula:C₁₇H₁₃ClNO₅P
:377.72 g/mol
Storage Temperature:Freezer
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together