ContactUsLinkComponent
Gel Cassette Kit, Sage Science
Gel Cassette Kit, Sage Science
Catalog # MSPP-HTG3010
Gel Cassette Kit, Sage Science
Catalog # MSPP-HTG3010
Supplier Number:  HTG3010
Specifications
  • Description:
    Gel cassette with 3% agarose
  • Size:
    12 samples/cassette
  • Cat. No.:
    MSPP-HTG3010
Specifications
About this item
Cassettes are disposable and pre-cast with agarose.
  • Uses marker 30G
  • For use with internal standards or external markers
Delivery: Package includes sufficient reagents for 10 cassettes.