Mouse Recombinant Recombinant Mouse R-Spondin 1 Protein, CF (from E. coli)
Supplier: Novus Biologicals
Certificates
About this item
R-spondin 1 (RSPO1, roof plate-specific Spondin 1), also known as cysteine-rich and single thrombospondin domain containing protein 3 (Cristin 3), is a 27 kDa secreted protein that belongs to the R-spondin family.
Specifications
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage Conditions:–20...–70 °C
- Gene ID:Rspo1
- Reconstitution Instructions:Reconstitute at 250 μg/ml in Sterile PBS
- Protein Synonyms:roof plate-specific spondin|Cristin 3|hRspo1|RSpondin 1|RSPO|FLJ40906Roof plate-specific spondin-1|RSPO1
- Protein/Peptide Name:Recombinant Mouse R-Spondin 1 Protein, CF
- Purity:>90%
- Endotoxin Level:<0.10 EU per 1 μg of the Protein by the LAL Method
- Formulation:Lyophilized from a 0.2 μm Filtered Solution in PBS
Specifications
Frequently Bought Together