ContactUsLinkComponent
Microcystins Strip Test Kit, Abraxis
Microcystins Strip Test Kit, Abraxis
  MSPP-520019
 :  
Microcystins Strip Test Kit, Abraxis
  MSPP-520019
 :  520019
 :  
  • Description:
    Microcystins Strip Kit
  • Cat. No.:
    MSPP-520019
  • No. of tests:
    5
The microcystins strip test for source drinking water is a rapid immunochromatographic test, designed solely for the use in the qualitative screening of microcystins and nodularins in source drinking water. A rapid cell lysis step (QuikLyse™) performed prior to testing is required to measure total microcystins (dissolved, or free, plus cell-bound). The microcystins strip test provides only preliminary qualitative test results. If necessary, positive samples can be confirmed by ELISA, HPLC or other conventional methods.
  • Store at 4 to 30 °C
 : Microcystins strip test kit contain microcystins test strips in a desiccated container, sample collection vials, lysis vials, graduated disposable pipettes (calibrated at 1 ml), forceps, reagent papers, conical test vials, disposable transfer pipettes and user’s guide.