Orders LinkContactUsLinkComponent
Microplate Vortex Mixers, Eberbach
Microplate Vortex Mixers, Eberbach
Supplier:   Eberbach
Microplate Vortex Mixers, Eberbach
Supplier:  Eberbach
Certificates
Microplate Vortex Mixers, Eberbach
2 Options Available Below
Product Details & Documents
These vortex lab shakers are specifically built for continuous duty and designed to produce vortex mixing of aqueous solutions in standard and deep well micro-titer plates.
  • Dynamically balanced large volume vortexing/single plane balanced small volume vortexing
  • Variable Speed digital display with digital countdown timer
  • Remote control operation
  • Tissue homogenization
  • OEM builds
  • Stainless steel models available
  • Incubator high humidity/CO₂ models
This unit has a digital programmable timer with a digital tachometer which allows timed protocols or the shaker may run continuously. The shaker has a rubber feet that prevent creeping.
Product Family Options
Product Information
  • Electrical
    Capacity
    Dimensions
    Orbit
    Speed
    Availability
    Price
  • 75836-358
    Vortex mixer, large
    Vortex mixer, large
    115/230 V, 50/60 Hz
    8 deep well titer plates or 16 standard plates
    14×14×6"
    0.125"
    120 - 2200 rpm
    Log in for availability
    Log in for availability
    Each
    Each - $8,319.23
    $8,319.23
    Each
       $8,319.23
    Minimum order quantity is 1
    Supplier Number: E6121.00
  • 75836-360
    Vortex mixer, large, stainless steel
    Vortex mixer, large, stainless steel
    115/230 V, 50/60 Hz
    8 deep well titer plates or 16 standard plates
    14×14×6"
    0.125"
    120 - 2200 rpm
    Log in for availability
    Log in for availability
    Each
    Each - $8,105.10
    $8,105.10
    Each
       $8,105.10
    Minimum order quantity is 1
    Supplier Number: E6121.00
  • 115/230 V, 50/60 Hz
    2 deep well titer plates or 4 standard plates
    7×6×6"
    0.125"
    120 - 2200 rpm
    Discontinued
    Supplier Number: -