Order Entry
United States
ContactUsLinkComponent
Radiance ECL, Azure Biosystems
Radiance ECL, Azure Biosystems
Catalog # 76318-638
Supplier:  Azure Biosystems
CAS Number:  
undefined
Radiance ECL, Azure Biosystems
Catalog # 76318-638
Supplier:  Azure Biosystems
Supplier Number:  AC2204
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Chemiluminescent HRP Substrate for Chemiluminescent Westerns
  • Size:
    500 ml
  • Cat. No.:
    76318-638

Specifications

About this item

Radiance ECL is an HRP substrate with long lasting signal and sensitivity down to low picograms. Use up to 10 times less antibody than other ECL substrates.

  • Compare performance to SuperSignal West Pico and Amersham ECL
  • Sensitivity in low picograms
  • Signal duration 6 to 8 hours