4'-Hydroxy-3'-methoxyacetophenone ≥98% (by GC), Calbiochem®, Sigma-Aldrich®
Catalog # 80512-324
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Plastic ampoule
- Size:1 g
- Cat. No.:80512-324
- Supplier no.:178385-1GM
Specifications
Specification Test Results
- Purity (by GC):≥98%
Specification Test Results
About this item
:
:166.18 g/mol
Merck Index:14,00741
:Packaged under inert gas.
Storage Temperature:Freezer
CAS Number:498-02-2
Formula:C₉H₁₀O₃
MDL Number:MFCD00008747